missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Isolator Pack Plated Media - Isolator 100mm TSA

Numéro de catalogue. B292652
Change view
Click to view available options
Certifications/Compliance:
AAMI
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Certifications/Compliance
B292652 AAMI
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. B292652 Fournisseur BD Code fournisseur 292652
Il en reste null

Plated media in gamma-irradiated packs for isolator systems

Plate types include Rodac™ (Replicate Organism Detection and Counting) for surface sampling and Finger Dab™ for sampling gloved hands.

To use:

  • Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
  • When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
  • Use the included inner bag to transport plates from your critical environment

Compliance

Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.

Spécifications

Certifications/Compliance AAMI
Color Red
Content And Storage 2°C to 8°C
Format 100 x 15mm Finger Dab
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar
Sterility Sterile
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Designed for isolator system
Includes Inner bag for transportation
Packaging Type Finger Dab™ Plate
Quantity 100/Pk.
Afficher plus de résultats Afficher moins de résultats