missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Urea Agar Base

Catalog No. B11795
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11795 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11795 Supplier BD Supplier No. 211795

For detection of urease producing bacteria

Specifications

Certifications/Compliance BAM, CCAM, ISO, USDA
Content And Storage Refrigerate
Form Dehydrated
Media Type Urea Agar Base
Product Type Agar
Formulation Pancreatic digest of Gelatin 1g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Potassium Phosphate 2g/L, Urea 20g/L, Phenol red 12mg/L
For Use With (Application) For differentiation of organisms, especially the Enterobacteriaceae, on the basis of Urease production
Packaging Type Bottle
Quantity 500 g