missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: Motility Test Medium

Catalog No. B11436
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11436 Supplier BD Supplier No. 211436
Edge

For demonstrating microorganism motility

Specifications

Certifications/Compliance BAM, CCAM, COMPF, USDA
Form Dehydrated
Media Type Motility Test Medium
Product Type Motility Test Medium
Formulation Beef extract 3g/L, Pancreatic digest of Casein 10g/L, Sodium Chloride 5g/L, Agar 4g/L
For Use With (Application) Demonstrating microorganisms motility
Packaging Type Bottle
Quantity 500 g