missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: MacConkey Agar
Spécifications
Spécifications
| Certifications/Compliance | ISO, 9000, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP |
| Content And Storage | 2°C to 30°C |
| Form | Dehydrated |
| Media Type | MacConkey Agar |
| Product Type | MacConkey Agar |
| Formulation | Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L |
| For Use With (Application) | Isolation of Enterococcus faecalis 29212, Escherichia coli 25922, Proteus mirabilis 12453, Salmonella enterica subspecies enterica serotype Typhimurium 14028, Shigella flexneri 12022, Escherichia coli 8739 |
| Packaging Type | Bottle |
| Quantity | 500 g |