missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Demi-Fraser Broth Base
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Demi-Fraser Broth Base |
| Product Type | Microbiology Media |
| Formulation | Tryptose 10g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.01g/L, Acriflavine HCl 12.5mg/L, Lithium Chloride 3g/L |
| For Use With (Application) | For use with Fraser broth supplement in selectively and differentially enriching Listeria, reduced Acriflavine and Nalidixic Acid levels |
| Packaging Type | Bottle |
| Quantity | 500 g |