missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bcl-W Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PA5150292
Modifier l'affichage
Click to view available options
Quantité:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PA5150292 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PA5150292 Fournisseur Invitrogen™ Code fournisseur PA5150292

Rabbit Polyclonal Antibody

Bcl-W Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Using a PCR-based strategy, an additional protein with life-promoting activity, designated Bcl-w, has been identified. The protein is highly conserved between mouse and human and is encoded by a gene located near the TCRalpha gene on chromosome 14. Bcl-w is expressed in myeloid cell lines but not in T and B lymphocytes, and can be found in a wide range of tissues. An alternative splicing event in exon 4 results in two transcripts. The first, Bcl-w, encodes a protein of 193 amino acids, and the second, Bcl-w/rox, encodes a protein 333 amino acids in length. The 'rox' portion of Bcl-w/rox shows a striking 66% amino acid sequence identity with the Drosophila rox2 protein; however, the Bcl-w/rox transcript may be expressed at very low levels.

Spécifications

Antigène Bcl-W
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gène BCL2L2
Numéro d’acquisition du gène P70345, Q92843
Alias du gène Apoptosis regulator Bcl-W; AW048834; BCL2 like 2; BCL2L2; BCL2-L-2; BCL2-like 2; BCL2-like 2 protein; Bcl-2-like protein 2; Bcl2-like protein 2; BCLW; BCL-W; BCL-WEL; BCL-WS; c98; Gtrgal2; Gtrosa41; Kiaa0271; PPP1R51; protein phosphatase 1, regulatory subunit 51
Symboles de gène BCL2L2
Espèces hôtes Rabbit
Immunogène Recombinant B-Cell CLL/Lymphoma 2 Like Protein 2 (Bcl2L2) (Ala2-Lys193).
Méthode de purification Protein A
Quantité 200 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 12050, 599, 60434
Espèces cibles Human, Mouse, Rat, Pig
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Afficher plus de résultats Afficher moins de résultats