missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Streptocard™ Enzyme Latex Test Kit

Catalog No. B240918
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B240918 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B240918 Supplier BD Supplier No. 240918

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

BBL Streptocard Enzyme Test Latex A, B, C, D, F and G: 2.5mL each; BBL Streptocard Enzyme Positive Control: 2.5mL; BD BBL Streptocard Extraction Enzyme: 22mL; Test cards; Mixing sticks

Provides a rapid platform for the serological identification of beta-haemolytic streptococci belonging to Lancefield groups A, B, C, D, F and G

Studies have shown that the diagnosis of streptococcal infections based on clinical symptoms requires microbiological verification. Beta- haemolytic streptococci are the most frequently isolated human pathogens among the representatives of the genus Streptococcus Nearly all the beta-haemolytic streptococci possess specific carbohydrate antigens (streptococcal group antigens). Lancefield showed that these antigens can be extracted in soluble form and identified by precipitation reactions with homologous antisera. The BD BBL Streptocard Enzyme Latex Test Kit is based on liberation of specific antigen from bacteria cell walls by the action of lytic enzymes. The extracted antigen in conjunction with latex agglutination offers a rapid, sensitive and specific method for identification of streptococcal groups A, B, C, D, F and G from primary culture plates.

Specifications

Content And Storage 2°C to 8°C Do not freeze
Description BBL Streptocard Enzyme Latex Test Kit
Detectable Analytes beta-haemolytic Streptococci (Groups A, B, C, F, G)
For Use With (Application) Streptococcus Testing
Includes BBL Streptocard Enzyme Test Latex A, B, C, D, F and G: 2.5mL each; BBL Streptocard Enzyme Positive Control: 2.5mL; BD BBL Streptocard Extraction Enzyme: 22mL; Test cards; Mixing sticks
Quantity 50 Tests