missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Media: CDC Anaerobe 5% Sheep Blood Agar

Catalog No. B21734X
Change view
Click to view available options
Quantity:
100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21734X 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21734X Supplier BD Supplier No. 221734
Only null left

For the isolation and cultivation of fastidious and slow growing, obligately anaerobic bacteria from a variety of clinical and nonclinical materials

It also supports the growth of most aerobic, facultatively anaerobic and microaerophilic bacteria if incubated appropriately.

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type CDC anaerobe 5% Sheep Blood Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 20g/L, Yeast extract 5g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, L-Cystine 0.4g/L, Sheep Blood, defibrinated 5%
For Use With (Application) Isolation and Cultivation of obligate anaerobes from clinical specimens
Packaging Type Plate
Quantity 100/Pk.
Weight (Metric) 950 g