missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ MycoPrep™ Mycobacterial System Digestion/Decontamination Kits

Catalog No. B4340862
Change view
Click to view available options
Volume (Metric):
75 mL
150 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
B4340862 75 mL
B4340863 150 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4340862 Supplier BD Supplier No. 240862

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

No weighing and no messy mixing

  • Provide one of the most widely used, CDC-recommended formulations for effective mycobacterial specimen processing
  • Convenient, all-inclusive kits are an alternative to traditional methods of preparing specimen digestion/decontamination reagents
  • Complete with bottles of NALC-NaOH Solution and pre-measured packets of Phosphate Buffer (pH 6.8)
  • Ensure standardization by eliminating the need to weigh, reconstitute or inventory separate ingredients
  • Remain stable for up to 24 hours after activation

Specifications

Product Type Specimen Collection Kit
Certifications/Compliance CE
For Use With (Application) For the digestion and decontamination of clinical specimens suspected to contain Mycobacteria, especially Mycobacterium tuberculosis
Includes 10 x 75 mL bottles of NALC-NaOH solution and 5 packets Phosphate Buffer (ph 6.8)
Quantity 10 x 75 mL Bottles
Volume (Metric) 75 mL
Storage Requirements 15°C to 25°C