missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ CultureSwab EZ™ and CultureSwab EZ II™

Catalog No. B220144
Change view
Click to view available options
Swab Configuration:
Dual Applicator
Single Applicator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Swab Configuration
B220144 Single Applicator
B220145 Dual Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B220144 Supplier BD Supplier No. 220144

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Polyurethane sponge swab on a plastic shaft secured to a cap, which is inserted into supplied plastic transport tube

A patented media-free collection system designed to collect aerobic specimens and maintain viability during transport

  • No specimen dilution with this dry system and no ampule to crush
  • Transport system eliminates the organism overgrowth and nonviables typically present in agar-based systems
  • Aerobic microorganisms remain viable for approximately 48 hours
  • Features rounded tube bottom for easier handling, and water-resistant film packing with tamper-evident seal
  • BD CultureSwab EZ II is similar to BD CultureSwab EZ except formatted as a dual swab system, allowing for simultaneous gram staining and other direct antigen testing

Specifications

Product Type Culture Swab™ EZ System
Sterility Sterile
Swab Configuration Single Applicator
For Use With (Application) For collecting and transporting aerobic microbiological specimens
Includes Polyurethane single sponge swab, Plastic shaft, cap, inserted into Plastic transport tube
Quantity 100/Pk.
Shaft Material Plastic
Storage Requirements 15°C to 30°C