missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BAY 60-6583

Catalog No. 447250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
447250 50 mg
447210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 447250 Supplier Tocris Bioscience™ Supplier No. 4472/50
Only null left

Potent A2B receptor agonist; cardioprotective

Potent adenosine A2B receptor agonist (EC50 = 2.83 nM for murine A2B receptor). Displays selectivity for A2B over A1, A2A and A3 receptors. Decreases fMLP-induced superoxide production in neutrophils at low concentrations (1-10 nM). Cardioprotective; attenuates infarct size in a mouse model of myocardial ischemia. Exhibits ERK1/2-biased agonism.

Chemical Identifiers

CAS 910487-58-0
Molecular Formula C19H17N5O2S

Specifications

Quantity 50 mg
Formula Weight 379.44
Purity >98%
Molecular Weight (g/mol) 379.438
Chemical Name or Material BAY 60-6583
Recommended Storage Store at +4°C