accessibility menu, dialog, popup

UserName

ONNSONH2H2NN

CAS RN 910487-58-0

IUPAC Name: 2-({6-amino-3,5-dicyano-4-[4-(cyclopropylmethoxy)phenyl]pyridin-2-yl}sulfanyl)acetamide
Synonyms: 2-({6-amino-3,5-dicyano-4-[4-(cyclopropylmethoxy)phenyl]pyridin-2-yl}sulfanyl)acetamide
Molecular Weight (g/mol): 379.44
Molecular Formula: C19H17N5O2S
InChi Key: ZTYHZMAZUWOXNC-UHFFFAOYSA-N
SMILES: NC(=O)CSC1=C(C#N)C(C2=CC=C(OCC3CC3)C=C2)=C(C#N)C(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results