missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAY 60-6583
Description
Potent adenosine A2B receptor agonist (EC50 = 2.83 nM for murine A2B receptor). Displays selectivity for A2B over A1, A2A and A3 receptors. Decreases fMLP-induced superoxide production in neutrophils at low concentrations (1-10 nM). Cardioprotective; attenuates infarct size in a mouse model of myocardial ischemia. Exhibits ERK1/2-biased agonism.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 379.44 |
| Purity | >98% |
| Molecular Weight (g/mol) | 379.438 |
| Chemical Name or Material | BAY 60-6583 |
Safety and Handling
| Recommended Storage | Store at +4°C |