missing translation for 'onlineSavingsMsg'
Learn More

BAY 11-7085 97.0+%, TCI America™

Catalog No. B484510MG
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B484510MG 10 mg
B4845100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B484510MG Supplier TCI America Supplier No. B484510MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 196309-76-9
Molecular Formula C13H15NO2S
Molecular Weight (g/mol) 249.328
MDL Number MFCD01862602
InChI Key VHKZGNPOHPFPER-ONNFQVAWSA-N
Synonym e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile
PubChem CID 5353432
IUPAC Name (E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile
SMILES CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N

Specifications

Melting Point 84°C
Color White
Quantity 10 mg
Formula Weight 249.33
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material BAY 11-7085
RTECSNumber UD1312500
TSCA No
Recommended Storage Refrigerator