missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Battery Cover for M210 Handheld Label Maker

Catalog No. 19000752
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19000752 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19000752 Supplier Brady™ Supplier No. M210BATTCOV

This is a replacement battery cover for the M210 Label Printer.

  • This is a replacement battery cover for the M210 Label Printer.
  • Package includes printer battery cover (printer and batteries sold separately).
  • Plastic cover snaps in place over the battery compartment.
  • Ensures your batteries remain securely in place.

Specifications

Product Type Printer
Color Yellow
Display Type None
For Use With (Equipment) Brady Labels, BMP21-PLUS Label Printer
Material Plastic
Width (English) 4.05 in.
Sterility Non-sterile