missing translation for 'onlineSavingsMsg'
Learn More

Bathophénanthroline 99,0+%, TCI America™

Numéro de catalogue. D09051G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D09051G 1 g
D09055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D09051G Fournisseur TCI America Code fournisseur D09051G

Identifiants chimiques

CAS 1662-01-7
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.406
MDL Number MFCD00004976
InChI Key DHDHJYNTEFLIHY-UHFFFAOYSA-N
Synonym bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
PubChem CID 72812
ChEBI CHEBI:77995
IUPAC Name 4,7-diphényl-1,10-phénanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5

Spécifications

Melting Point 221°C
Color Blanc-jaune
Quantity 1 g
Formula Weight 332.41
Percent Purity ≥99.0% (T)
Physical Form Poudre cristalline
Chemical Name or Material Bathophenanthroline
RTECSNumber SF8427000
TSCA Yes