Learn More
Invitrogen™ BATF Monoclonal Antibody (MBM7C7), PerCP-eFluor™ 710, eBioscience™

Description
Description: The MBM7C7 monoclonal antibody reacts with human and mouse BATF (B cell activating transcription factor). BATF is a 14 kDa member of the AP-1 family of transcription factors and is involved in the repression of other AP-1 family members. BATF is upregulated in activated T cells when compared to naive T and B cells and has been shown to be critical for Th17, but not Th1 or Th2 differentiation. In Th17, BATF interacts with the promoters of IL17, IL17F, IL21 and IL22. BATF has also been shown to be involved in CD8 T cell exhaustion in HIV infection. Applications Reported: This MBM7C7 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MBM7C7 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated human peripheral blood cells using the Foxp3/Transcription Factor Buffer Set (Product # 00-5523-00) and protocol. Please see BestProtocols® Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be...
Spécifications
Spécifications
| Antigène | BATF |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MBM7C7 |
| Concentration | 5 μL/Test |
| Conjugué | PerCP-eFluor 710 |
| Formule | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gène | Batf |
| Numéro d’acquisition du gène | Q16520 |
| Alias du gène | activating transcription factor B; basic leucine zipper ATF-like transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; basic leucine zipper transcriptional factor ATF-like; BATF; B-ATF; BATF1; B-cell activating transcription factor; B-cell-activating transcription factor; SFA2; SFA-2; SF-HT-activated gene 2 protein; si:ch211-153j24.4 |
| Afficher plus de résultats |