missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Basic Yellow 1 95.0+%, TCI America™
Description
Basic Yellow 1 (Setoflavine T, Thioflavine T) is a laboratory reagent supplied as a solid powder or crystal with a purity of ≥95.0% (HPLC).
- Molecular Formula: C17H19ClN2S
- Molecular Weight: 318.86 g/mol
- Melting Point: 212 °C
- Water Solubility: 17.6 g/L (20 °C)
- Optical Properties: λmax 410.0 to 414.0 nm (H2O); Absorbance (E1%1cm) ≥750
- Safety Information: Toxic if swallowed (H301); causes serious eye damage (H318); may cause an allergic skin reaction (H317); very toxic to aquatic life with long lasting effects (H410)
Specifications
Specifications
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 318.86 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Laboratory |
| Chemical Name or Material | Basic Yellow 1 |
Safety and Handling
| EINECSNumber | (5)-1916 |
| TSCA | Yes |