missing translation for 'onlineSavingsMsg'
Learn More

Basic Fuchsin, Spectrum™ Chemical

Catalog No. 18600856
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600856 1 kg Amber Glass Bottle
18600853 5 g Amber Glass Bottle
18600854 25 g Amber Glass Bottle
18600855 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600856 Supplier Spectrum Chemical Mfg Cor Supplier No. BA1301KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BA130, 569-61-9, C19H17N3·HCl

Spectrum™ Chemical Basic Fuchsin is a magenta dye that is a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin is a dark green crystal often used in the laboratory where it is employed to stain bacteria and in the fabrication of Schiff reagent. It is made up of a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin also has uses in the textile industry, and has many other names, including Basic Parafuchsin, Basic Red 9, Paramagenta Hydrochloride, and Pararosaniline Chloride.

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.82
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
IUPAC Name hydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
SMILES [H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Grade Reagent
Physical Form Crystalline Powder or Lumps