missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Basic Fuchsin, 70+%, pure
CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol
$617.51 - $617.51
Identifiants chimiques
| CAS | 632-99-5 |
|---|---|
| Molecular Formula | C20H20ClN3 |
| Molecular Weight (g/mol) | 337.85 |
| MDL Number | MFCD00012569 |
| InChI Key | AXDJCCTWPBKUKL-UHFFFAOYSA-N |
| Synonym | Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride |
| PubChem CID | 12447 |
| IUPAC Name | 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
| SMILES | [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC401691000
|
Thermo Scientific Chemicals
401691000 |
100 g | Glass bottle |
chaque for $617.51
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 632-99-5 | |
| 337.85 | |
| AXDJCCTWPBKUKL-UHFFFAOYSA-N | |
| 12447 | |
| [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1 |
| C20H20ClN3 | |
| MFCD00012569 | |
| Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride | |
| 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
Spécifications
| 250.0°C | |
| 1.16 to 1.35 | |
| C20H20ClN3 | |
| 13, 765 | |
| Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride | |
| AXDJCCTWPBKUKL-UHFFFAOYSA-N | |
| [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1 | |
| 337.85 | |
| 547 to 552nm | |
| ≥70% | |
| Glass bottle | |
| Green | |
| 200°C | |
| Basic Fuchsin, Pure |
| 632-99-5 | |
| 70% min. | |
| MFCD00012569 | |
| 15, 5715 | |
| Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C) | |
| Authentic | |
| 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride | |
| 12447 | |
| 337.84 | |
| Pure | |
| 200g/mL | |
| 100 g | |
| Glistening Crystalline Powder |
Sécurité et manipulation
GHS H Statement
Suspected of causing cancer. Harmful if swallowed.
GHS P Statement
Use personal protective equipment as required. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Warning
RUO – Research Use Only