missing translation for 'onlineSavingsMsg'
Learn More

Basic Blue 7, TCI America™

Numéro de catalogue. B130125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B130125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. B130125G Fournisseur TCI America Code fournisseur B130125G
Il en reste null

Identifiants chimiques

CAS 2390-60-5
Molecular Formula C33H40ClN3
Molecular Weight (g/mol) 514.15
MDL Number MFCD00011877
InChI Key ZQTYUKTXBQYBSD-UHFFFAOYSA-M
Synonym Victoria Pure Blue BO
PubChem CID 75444
IUPAC Name bis[4-(diethylamino)phenyl][4-(ethylamino)naphthalen-1-yl]methylium chloride
SMILES [Cl-].CCNC1=C2C=CC=CC2=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC

Spécifications

Formula Weight 514.15
Color Red/Yellow
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Basic Blue 7
EINECSNumber (5)-1994
TSCA Yes