missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Base Retort Stand

Rectangular in shape.

Supplier:  Eisco™ CH0652B

 Afficher plus de versions de ce produit

Numéro de catalogue. S02611


Il en reste null

Description

Description

  • Pressed steel base
  • With hole 10 x1.5mm metric thread in the center of one short side
  • Finished in blue/grey colour
Spécifications

Identifiants chimiques

Steel
Rectangular
Blue/Gray
Base Retort Stand
0.39 x 0.05 in.
Each

Spécifications

Steel
1.1 in.
20 cm
12.5 cm
Rectangular
Blue/Gray
3.6 lb.
3 cm
7.8 in.
4.9 in.
Base Retort Stand
0.39 x 0.05 in.
Each
1.6 kg
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions