missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Base/Neutrals Compounds 2B - WP, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-64-1 |
| Flash Point | -17°C - closed cup - Solvent |
| Type | Proficiency Testing Material |
| Quantity | 2 mL |