missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, Powder, Spectrum™ Chemical

Catalog No. 18609030
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609030 25 kg Poly Bottle
18609026 125 g Poly Bottle
18609027 500 g Poly Bottle
18609029 2.5 kg Poly Bottle
18609028 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609030 Supplier Spectrum Chemical Mfg Cor Supplier No. B105525KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7727-43-7 | 233.39 g/mol

Chemical Identifiers

CAS 7727-43-7

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 25 kg
Molecular Weight (g/mol) 233.39
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Barium Sulfate, Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.