missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Sulfate, Multi-Compendial, U.S.P., J.T. Baker™
Specifications
Specifications
| Quantity | 500 g |
| Formula Weight | 233.39 |
| Percent Purity | 97.7% to 100.5% |
| Grade | Multi-Compendial/USP |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 4.50kg |
| Chemical Name or Material | Barium Sulfate |