missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfate, 98%, extra pure

Catalog No. AC222515000
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222515000 500 g Plastic bottle
AC222510010 1 kg Plastic bottle
AC222510050 5 kg Plastic bottle
AC222510250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC222515000 Supplier Thermo Scientific Chemicals Supplier No. 222515000

Barium sulfate, 98%, BaO4S, CAS Number-7727-43-7, barite, baritop, barium sulfate, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast, 25kg, Passes test, < 0.3%, 97.5 to 100.5%, White, 4.5000g/mL, 231-784-4, 233.4, Extra Pure, 10ppm max., TZCXTZWJZNENPQ-UHFFFAOYSA-L | CAS: 7727-43-7 | BaO4S | 233.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 97.5 to 100.5%
Linear Formula BaSO4
Merck Index 15,99
Solubility Information Solubility in water: 0.0022g/L (18°C). Other solubilities: practically insoluble in diluted acids, soluble in hot conc. sulfuric acid, insoluble in ethanol
IUPAC Name barium(2+);sulfate
Formula Weight 233.4
Percent Purity 98%
Grade Extra Pure
Loss on Ignition 2.0% (600°C) max.
Packaging Plastic bottle
Specific Gravity 4.5
Density 4.5000g/mL
Chemical Name or Material Barium sulfate
Show More Show Less
EINECSNumber 231-784-4

RUO – Research Use Only