missing translation for 'onlineSavingsMsg'
Learn More

Barium manganate, tech. 90%

Catalog No. AAL1317214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1317214 25 g
AAL1317206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1317214 Supplier Thermo Scientific Chemicals Supplier No. L1317214
Only null left

CAS: 7787-35-1 | BaMnO4 | 256.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-35-1
Molecular Formula BaMnO4
Molecular Weight (g/mol) 256.26
MDL Number MFCD00011514
InChI Key ZZCNKSMCIZCVDR-UHFFFAOYSA-N
Synonym barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese
PubChem CID 3084030
IUPAC Name barium(2+);dioxido(dioxo)manganese
SMILES [Ba++].[O-][Mn]([O-])(=O)=O

Specifications

Quantity 25 g
UN Number UN3087
Merck Index 14,981
Formula Weight 256.28
Percent Purity 90%
Grade Technical
Sensitivity Moisture sensitive
Density 4.85
Chemical Name or Material Barium manganate
Hazard Category H272-H302+H332
Hazard Statement GHS H Statement
H272-H331-H302
May intensify fire; oxidizer.
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
EINECSNumber 232-109-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only