missing translation for 'onlineSavingsMsg'
Learn More

Barium ICP Standard, 1000 ppm Ba in 3% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company PBA1KH500

Catalog No. PBA1KH500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Barium ICP Standard
513-77-9 , 7647-01-0 , 7732-18-5
1000ppm Ba
1 mL = 1 mg Ba
MFCD00003448
[Ba++].[O-]C([O-])=O
197.34
Natural Poly Bottle
94.0,3.48,0.14
98.0,3.62,0.14
CBaO3
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+) carbonate
10563

Chemical Identifiers

513-77-9
197.34
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+) carbonate
CBaO3
MFCD00003448
10563
[Ba++].[O-]C([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions