accessibility menu, dialog, popup

UserName

BaOOO

CAS RN 513-77-9

IUPAC Name: barium(2+) carbonate
Synonyms: barium carbonate
Molecular Weight (g/mol): 197.34
Molecular Formula: CBaO3
InChi Key: AYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES: [Ba++].[O-]C([O-])=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (7)
Physical Form 
  • (4)
  • (4)
  • (16)
  • (5)
Grade 
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (9)
  • (1)
  • (1)
  • (9)
  • (1)
  • (22)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (7)
  • (4)
  • (4)
  • (16)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)