missing translation for 'onlineSavingsMsg'
Learn More

Barium Diphenylamine Sulfonate, Reagents

Catalog No. 01337471
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337471 25 g
01337470 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337471 Supplier Labchem Inc Supplier No. C214725025C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Barium Diphenylamine Sulfonate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym Barium diphenylamine-4-sulfonate, N-Phenylsulfanilic Acid Barium Salt
IUPAC Name barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color White to Gray
Physical Form Solid
Quantity 25 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0001
Chemical Name or Material Barium Diphenylamine Sulfonate