missing translation for 'onlineSavingsMsg'
Learn More

Barium Diphenylamine Sulfonate, Reagent Grade, LabChem™

Catalog No. LC116258
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC116258 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC116258 Supplier LabChem Supplier No. LC116258

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity chemical reagent for laboratory and manufacturing applications.

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym barium diphenylamine-4-sulfonate, barium diphenylaminesulfonate, benzenesulfonic acid, 4-phenylamino-, barium salt 2:1, barium bis 4-anilinobenzenesulphonate, barium diphenylamine sulfonate, n-phenylsufanilic acid barium, unii-ssj5f79159, diphenylamine-4-sulfonic acid barium salt, n-phenylsufanilic acid, barium salt, 4-anilinobenzenesulfonate; barium 2+
PubChem CID 80342
IUPAC Name barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 300°C
Color White
CAS Max % 100
Physical Form Powder
Quantity 25 g
Linear Formula C24H20N2S2O6Ba
Solubility Information Soluble in water
Formula Weight 633.9
Grade Reagent
Identification Passes Test
Packaging Amber Glass
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Barium; Nitrogen oxides
Chemical Name or Material Barium Diphenylamine Sulfonate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed or if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 228-278-0
Recommended Storage Room Temperature