missing translation for 'onlineSavingsMsg'
Learn More

Barium diphenylamine-4-sulfonate

Catalog No. AAA1619714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1619714 25 g
AAA1619706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1619714 Supplier Thermo Scientific Chemicals Supplier No. A1619714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6211-24-1 | C24H20BaN2O6S2 | 633.88 g/mol

Barium diphenylamine-4-sulfonate is used as a pharmaceutical intermediate and in chemical research. It is also used as an analytical dosing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium diphenylamine-4-sulfonate is used as a pharmaceutical intermediate and in chemical research. It is also used as an analytical dosing agent.

Solubility
It is moderate soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with mineral acids, organic acids, strong oxidizing agents.

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym barium diphenylamine-4-sulfonate, barium diphenylaminesulfonate, benzenesulfonic acid, 4-phenylamino-, barium salt 2:1, barium bis 4-anilinobenzenesulphonate, barium diphenylamine sulfonate, n-phenylsufanilic acid barium, unii-ssj5f79159, diphenylamine-4-sulfonic acid barium salt, n-phenylsufanilic acid, barium salt, 4-anilinobenzenesulfonate; barium 2+
PubChem CID 80342
IUPAC Name 4-anilinobenzenesulfonate;barium(2+)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point >300°C
Quantity 25 g
UN Number UN1564
Solubility Information It is moderate soluble in water.
Formula Weight 633.9
Chemical Name or Material Barium diphenylamine-4-sulfonate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 228-278-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only