missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride TS/RS, 12% (w/v), Ricca Chemical

Catalog No. R085400020F
Change view
Click to view available options
Package: Cubitainer; Size: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R085400020F Package: Cubitainer; Size: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R085400020F Supplier Ricca Chemical Company Supplier No. R085400020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 10326-27-9, 7732-18-5
CAS Min % 87.0, 10.51
CAS Max % 91.0, 10.93
Molecular Formula BaCl2H4O2
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES O.O.[Cl-].[Cl-].[Ba+2]
IUPAC Name barium(2+);dichloride;dihydrate
Molecular Weight (g/mol) 244.257
PubChem CID 5284346
ChEBI CHEBI:86153
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 20 L
Concentration or Composition Notes 12% (w/v)
Chemical Name or Material Barium Chloride, Test Solution/Reagent Solution
Show More Show Less