missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride solution, 0.1M, Solstice

Catalog No. 6002486
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002486 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002486 Supplier Solstice Supplier No. 342521L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Volumetric, 0.1 M BaCl2 (0.2N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+);dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Quantity 1 L
UN Number NONH for all modes of transport
Formula Weight 208.23g/mol
Concentration or Composition (by Analyte or Components) 0.1M
Chemical Name or Material Barium chloride solution