missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCB0080500
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCB0080500 500 g White Poly Wide Mouth Bottle
RDCB0080100 100 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCB0080500 Supplier Ricca Chemical Company Supplier No. RDCB0080500B1
Only null left

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals or Powder
Quantity 500 g
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
Percent Purity ≥99%
Grade ACS
Loss on Drying 14.0 to 16.0%
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Barium Chloride, Dihydrate
Show More Show Less
ShelfLife 60 months