missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 22.5°C Baumé, Ricca Chemical

Catalog No. p-4773736
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R0870000500 500 mL Natural Poly Bottle
R08700004A 4 L Natural Poly Bottle
R087000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R0870000500 Supplier Ricca Chemical Company Supplier No. R0870000500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10326-27-9 , 7732-18-5
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.257
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Ba+2]

Specifications

Packaging Natural Poly Bottle
Quantity 500 mL
Chemical Name or Material Barium Chloride, 22.5°F Baumæ at 60°
ShelfLife 24 months