missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 99+% |
| Linear Formula | BaCO3 |
| Merck Index | 15, 965 |
| IUPAC Name | barium(2+);carbonate |
| Formula Weight | 197.35 |
| Percent Purity | 99.0 to 101.0% |
| Grade | ACS Reagent |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | Warning |
| missing translation for 'healthHazard2' | GHS H Statement: Harmful if swallowed. |
| missing translation for 'healthHazard3' | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 208-167-3 |
| RTECSNumber | CQ8600000 |
| TSCA | TSCA |
RUO – Research Use Only