missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide dihydrate, 98+%

Catalog No. AAA1632836
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1632836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1632836 Supplier Thermo Scientific Chemicals Supplier No. A1632836
Only null left

CAS: 7791-28-8 | BaBr2H4O2 | 333.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-28-8
Molecular Formula BaBr2H4O2
Molecular Weight (g/mol) 333.17
MDL Number MFCD00149153
InChI Key HQQMRSGBIPFNSN-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dibromide
SMILES O.O.[Br-].[Br-].[Ba++]

Specifications

Quantity 500 g
UN Number UN1564
Merck Index 14,968
Formula Weight 333.17 (297.15 Anhydrous)
Percent Purity ≥98%
Sensitivity Hygroscopic
Density 3.58
Chemical Name or Material Barium bromide dihydrate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 234-140-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only