missing translation for 'onlineSavingsMsg'
Learn More

Barium AA Standard, Certified, 1000ppm Ba ±10ppm (1mL = 1mg Ba), LabChem™

Catalog No. LC115507
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC115507 125 mL Poly Bottle
LC115501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC115507 Supplier LabChem Supplier No. LC115507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10361-37-2 , 7732-18-5
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+) dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Packaging Poly Bottle
CAS Max % 99.82, 0.18
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm Ba ±10 ppm (1mL = 1mg Ba)
Linear Formula BaCl2·2H2O
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 244.26
Grade Certified
Chemical Name or Material Barium AA Standard, 1000ppm (1mL = 1mg Ba)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature