missing translation for 'onlineSavingsMsg'
Learn More

Barium AA Standard, 1000 ppm Ba in 3% HCl, Ricca Chemical

Catalog No. p-3251331
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
ABA1KH100 Natural LDPE Bottle
ABA1KH500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABA1KH100 Supplier Ricca Chemical Company Supplier No. ABA1KH100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10361-37-2 , 7647-01-0 , 7732-18-5
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.227
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+);dichloride
SMILES [Cl-].[Cl-].[Ba+2]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Ba
CAS Min % 94.0, 3.45, 0.15
CAS Max % 98.0, 3.59, 0.15
Equivalence 1 mL = 1 mg Ba
Chemical Name or Material Barium AA Standard