Learn More
Avantor™ BAKERBOND Speedisk™ Octadecyl (C18) Extraction Disk
Catalog No. 02002376
Change view
Click to view available options
Sorbent:
C18 (high capacity)
Catalog No. Sorbent
02002376 C18 (high capacity)
1 options
Catalog No. 02002376 Supplier Avantor™ Supplier No. 805507
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Octadecyl (C18) Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
For Use With (Application) EPA Methods 500 Series, 608 SW 846/3535 and with Slightly Polar to Non-Polar Industrial Samples
For Use With (Equipment) Expanded Extraction Station (8095-06)
Sorbent C18 (high capacity)
Type Extraction Disk
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm