Learn More
Research Products Corporation Electronic Bag Sealer, 12 Inch Heating Surface
Numéro de catalogue. 501367721
Encompass
Convenient Electronic Bag Sealer is perfect for sealing all polyethylene bags quickly. Easy access control knob allows for quick temperature adjustments. Specially designed heating element is hot only when sealing pressure is applied.
Heat Sealable Poly Bags for use with Bag Sealer are also available from RPI. Optional accessories for bag sealer replacement or repair are also available, sold separately.
Numéro de catalogue. 501367721 Fournisseur Research Products Corporation Code fournisseur 248101
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Convenient Electronic Bag Sealer is perfect for sealing all polyethylene bags quickly. Easy access control knob allows for quick temperature adjustments. Specially designed heating element is hot only when sealing pressure is applied.
Heat Sealable Poly Bags for use with Bag Sealer are also available from RPI. Optional accessories for bag sealer replacement or repair are also available, sold separately.