missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Proteose Peptone No. 2

Enzymatic digest of protein

Supplier:  Gibco™ 212120

 Afficher plus de versions de ce produit

Numéro de catalogue. DF0121-17-5


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Used in preparing microbiological culture media.
Spécifications

Spécifications

LUO
Mammalian, Insect, Microbial
Bacterial Media
Formulation of culture media for the cultivation of heterotrophic microorganisms
500 g
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Powder
Bottle
Bacto
Animal Origin
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions