missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Neopeptone
GREENER_CHOICE

An animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation.

$605.93 - $12,354.75

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
For Use With (Application) Production of vaccines for animal health applications
Product Line Bacto
Source Animal origin, bovine and porcine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
DF0119179
GREENER_CHOICE
View Documents Promotion Details
Gibco™
211681
500 g
Each for $605.93
Only null left
 
211680
GREENER_CHOICE
View Documents Promotion Details
Gibco™
211680
10 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein
  • This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation
  • It has been used as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci
  • Also used with Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health application
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Applications of Bacto Neopeptone include:

  • Recommended for use in media for detection of fungi
  • Reported to provide nutrients for support of spirochetes and protozoa
  • Microbial fermentation
  • Production of vaccines for animal health applications
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Production of vaccines for animal health applications
Animal origin, bovine and porcine
Powder
Bacto
Videos
Safety and Handling

Safety and Handling

ShelfLife : 1825 Days

SDS
Documents

Documents