missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Neopeptone
GREENER_CHOICE

Catalog No. DF0119179
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0119179 500 g
211680 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0119179 Supplier Gibco™ Supplier No. 211681

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation.

  • Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein
  • This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation
  • It has been used as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci
  • Also used with Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health application
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Applications of Bacto Neopeptone include:

  • Recommended for use in media for detection of fungi
  • Reported to provide nutrients for support of spirochetes and protozoa
  • Microbial fermentation
  • Production of vaccines for animal health applications

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Neopeptone
Format Bottle
For Use With (Application) Production of vaccines for animal health applications
Product Line Bacto
Quantity 500 g
Source Animal origin, bovine and porcine