missing translation for 'onlineSavingsMsg'
Learn More

BD BACTEC™ Standard/10 Aerobic/F Culture Vials

Catalog No. B442027
Change view
Click to view available options
Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
B442027 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B442027 Supplier BD Supplier No. B442027

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A complete line of blood culture media for the detection of aerobes, anaerobes, facultative anaerobes, yeast, fungi and mycobacteria, all specifically developed to help improve time of detection and organism recovery.

  • BD BACTEC™ Standard Anaerobic medium is for the qualitative culture and recovery of anaerobic microorganisms (bacteria and yeast) from blood.

Specifications

Product Type Blood Culture Media Vial
Target Organism Class Aerobic Microorganisms
Test Type Microbial Detection
Species Human