missing translation for 'onlineSavingsMsg'
Learn More

Decon™ BDD™ Bacdown™ Detergent Disinfectant
GREENER_CHOICE

Catalog No. 18800101
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
18800101 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Corrosive - chemical will cause eye damage and skin burns. Harmful or fatal if swallowed. Hazardous to aquatic environments.
Catalog No. 18800101 Supplier Decon™ Supplier No. DC8004
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Corrosive - chemical will cause eye damage and skin burns. Harmful or fatal if swallowed. Hazardous to aquatic environments.

Bacdown™ concentrated quaternary disinfectant cleans and disinfects surfaces with biological decontamination against HIV-1, HBV and others.

BDD™ (Bacdown™ Detergent Disinfectant) is a concentrated, phosphate-free, disinfectant containing surfactants and quaternary ammonium compounds that will safely and effectively clean and disinfect surfaces in labs, production areas and process equipment. Bactericidal, virucidal, fungicidal, and provides effective biological decontamination against HIV-1, HBV and more when used as instructed. Especially formulated for cleaning and sanitizing inanimate hard, non-porous surfaces.

Compliance

OSHA Bloodborne Pathogens Standards, EPA registered

Specifications

Product Type Disinfectant Cleaner
Color-coded No
Form Concentrated Liquid
Scent Regular Scent
Sterility Non-sterile
Appearance Clear Pale Yellow
Certifications/Compliance OSHA Bloodborne Pathogens Standard; EPA registered
Composition 2.25% N-alkyl Dimethyl Benzyl Ammonium Chlorides, 2.25% Ammonium Chlorides
Container Type Plastic Container
Content And Storage Store in Dry Place Between 50°F and 120°F
Dilution 2 oz./gal. of water (1:64)
Disposable Pesticide disposal. Do not reuse container. Triple rinse before recycling
Length (Metric) 30.48 cm
Quantity 4/Cs.
Size 1 g
Volume (English) 1 gal.
Width (Metric) 30.48 cm
Show More Show Less