Learn More
ACROBiosystems Inc Recombinant Protein;Biotinylated Human B7-H3 / CD276 Protein, His Tag & Avi Tag (Avitag™);HEK293;200UG;B73,B7-H3,CD276,B7 homolog 3,B7H3
Numéro de catalogue. 501658282
Encompass
Recombinant Protein;Biotinylated Human B7-H3 / CD276 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human B7-H3, His Tag (B73-H82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 26.5 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Numéro de catalogue. 501658282 Fournisseur ACROBiosystems Inc Code fournisseur B73H82E6200UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Biotinylated Human B7-H3 / CD276 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human B7-H3, His Tag (B73-H82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 26.5 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.