missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ B6-White™ Murine Embryonic Stem Cell Line

Catalog No. SCR011
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR011 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR011 Supplier MilliporeSigma™ Supplier No. SCR011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acts as fundamental tool for analyzing gene function

  • Strain: C57BL/6-tyr(c-2J) albino mice
  • Gender: Male
  • Karyotype: 40XY

Specifications

Content And Storage Place vials in the vapor phase of liquid nitrogen storage immediately upon receipt until it is convenient to proceed to subculture.
Cell Type Murine Embryonic Stem Cells
For Use With (Application) Stem cell culture
Quantity 2 Vials
Remarks Frozen in basal medium supplemented with 10% FCS and 10% DMSO

For Research Use Only