Learn More
Brady™ B30 Series Metallized Polyester Label Material
Designed to withstand numerous solvents and variable temperatures
$382.63 - $466.46
| Catalog Number | Mfr. No. | Adhesive Type | Finish | Thickness (English) | Temperature Range (English) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Adhesive Type | Finish | Thickness (English) | Temperature Range (English) | Price | Quantity & Availability | ||
|
19100027
|
Brady™
B30C4000428 |
Permanent | Matte | 0.0044 in. | -40° to +248°F |
Each for $443.48
|
|
||
|
19100029
|
Brady™
B30C4000434 |
Acrylic, Permanent | Gloss | 0.004 in. | -40° to +194°F |
Each for $382.63
|
|
||
|
19100040
|
Brady™
B30C4000486 |
Permanent | Matte | 0.0053 in. | -40° to +248°F |
Each for $466.46
|
|
||
Description
- Metallized polyester material
- Matte or glossy finish
- Permanent acrylic or rubber-based adhesive
- For applications like labelling rating and serial plates
Specifications
| Metallized Polyester |